Assay Details
| Gene Symbol: chp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059118.6 | NP_491519.1 | 2 - 3 | 1162 | 104 | |
| GenBank mRNA | AF192264.1 | - | 1 - 2 | 71 | 104 | |
| Gene Symbol: pas-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383045.1 | 2 - 3 | 1161 | 104 | ||
| Gene Symbol: spe-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059117.3 | NP_491518.1 | 1 - 2 | 72 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059117.3 transcripts.
Set Membership: |
|

chp-1
Genomic Map