Assay Details
| Gene Symbol: aap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059121.8 | NP_491522.1 | 1 - 2 | 182 | 130 | |
| GenBank mRNA | AF209707.1 | - | 1 - 2 | 171 | 130 | |
More Information
Set Membership: |
|

aap-1
Genomic Map