Assay Details
| Gene Symbol: aap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059121.8 | NP_491522.1 | 2 - 3 | 444 | 88 | |
| GenBank mRNA | AF209707.1 | - | 2 - 3 | 433 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aap-1| Gene Symbol: aap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059121.8 | NP_491522.1 | 2 - 3 | 444 | 88 | |
| GenBank mRNA | AF209707.1 | - | 2 - 3 | 433 | 88 | |
Set Membership: |
|