Assay Details
| Gene Symbol: mtm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059130.5 | NP_491531.2 | 8 - 9 | 1532 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtm-1| Gene Symbol: mtm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059130.5 | NP_491531.2 | 8 - 9 | 1532 | 108 | |
Set Membership: |
|