Assay Details
| Gene Symbol: osr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059144.6 | NP_491545.2 | 4 - 5 | 991 | 114 | |
| GenBank mRNA | AY360470.1 | - | 4 - 5 | 991 | 114 | |
More Information
Set Membership: |
|

osr-1
Genomic Map