Assay Details
| Gene Symbol: atp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026250.7 | NP_001021421.1 | 3 - 4 | 482 | 75 | |
| NM_001383096.1 | 3 - 4 | 468 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atp-3| Gene Symbol: atp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026250.7 | NP_001021421.1 | 3 - 4 | 482 | 75 | |
| NM_001383096.1 | 3 - 4 | 468 | 75 | |||
Set Membership: |
|