Assay Details
| Gene Symbol: F27C1.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374894.2 | NP_001362169.1 | 28 | 3789 | 98 | |
| NM_001374895.3 | NP_001362170.1 | 23 - 24 | 3327 | 98 | ||
More Information
Set Membership: |
|

F27C1.11
Genomic Map