Assay Details
| Gene Symbol: F27C1.13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059200.4 | NP_491601.4 | 4 - 5 | 331 | 76 | |
| Gene Symbol: F27C1.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374894.2 | NP_001362169.1 | 4 | 349 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001374894.2 transcripts.
Set Membership: |
|

F27C1.13
Genomic Map