Assay Details
| Gene Symbol: rpa-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383099.1 | 3 - 4 | 329 | 71 | ||
| NR_132558.1 | 3 - 4 | 309 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpa-4| Gene Symbol: rpa-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383099.1 | 3 - 4 | 329 | 71 | ||
| NR_132558.1 | 3 - 4 | 309 | 71 | |||
Set Membership: |
|