Assay Details
| Gene Symbol: cpd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059218.8 | NP_491619.2 | 20 - 21 | 2953 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpd-1| Gene Symbol: cpd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059218.8 | NP_491619.2 | 20 - 21 | 2953 | 109 | |
Set Membership: |
|