Assay Details
| Gene Symbol: tom-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129022.5 | NP_001122494.1 | 13 - 14 | 2112 | 91 | |
| NM_001129023.2 | NP_001122495.1 | 13 - 14 | 2115 | 91 | ||
| NM_059222.7 | NP_491623.3 | 13 - 14 | 2113 | 91 | ||
| GenBank mRNA | AY912102.1 | - | 13 - 14 | 2006 | 91 | |
| AY997305.1 | - | 13 - 14 | 2006 | 91 | ||
| AY997306.1 | - | 13 - 14 | 2006 | 91 | ||
More Information
Set Membership: |
|

tom-1
Genomic Map