Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 17 - 18 | 3080 | 144 | |
| NM_001318273.3 | NP_001305202.1 | 5 - 6 | 839 | 144 | ||
| NM_001318274.3 | NP_001305203.1 | 1 - 2 | 80 | 144 | ||
| NM_001383115.1 | 18 - 19 | 3164 | 144 | |||
| NM_059236.5 | NP_491637.2 | 19 - 20 | 3203 | 144 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map