Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 1 - 2 | 54 | 76 | |
| NM_001383115.1 | 2 - 3 | 138 | 76 | |||
| NM_059236.5 | NP_491637.2 | 3 - 4 | 177 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383115.1 transcripts.
Set Membership: |
|

tns-1
Genomic Map