Assay Details
| Gene Symbol: cir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059253.5 | NP_491654.1 | 3 - 4 | 999 | 79 | |
| GenBank mRNA | DQ645890.1 | - | 3 - 4 | 1021 | 79 | |
More Information
Set Membership: |
|

cir-1
Genomic Map