Assay Details
| Gene Symbol: cpb-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059279.7 | NP_491680.2 | 2 - 3 | 1083 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpb-3| Gene Symbol: cpb-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059279.7 | NP_491680.2 | 2 - 3 | 1083 | 106 | |
Set Membership: |
|