Assay Details
| Gene Symbol: glh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059280.5 | NP_491681.1 | 1 - 2 | 163 | 83 | |
| GenBank mRNA | AF079509.1 | - | 1 - 2 | 185 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

glh-3| Gene Symbol: glh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059280.5 | NP_491681.1 | 1 - 2 | 163 | 83 | |
| GenBank mRNA | AF079509.1 | - | 1 - 2 | 185 | 83 | |
Set Membership: |
|