Assay Details
| Gene Symbol: glh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059280.5 | NP_491681.1 | 3 - 4 | 1749 | 122 | |
| GenBank mRNA | AF079509.1 | - | 3 - 4 | 1771 | 122 | |
More Information
Set Membership: |
|

glh-3
Genomic Map