Assay Details
| Gene Symbol: pgp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313482.3 | NP_001300411.1 | 1 - 2 | 167 | 164 | |
| NM_001313483.4 | NP_001300412.1 | 1 - 2 | 243 | 164 | ||
More Information
Set Membership: |
|

pgp-2
Genomic Map