Assay Details
| Gene Symbol: pgp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313481.3 | NP_001300410.1 | 5 - 6 | 1025 | 109 | |
| NM_001313482.3 | NP_001300411.1 | 5 - 6 | 1124 | 109 | ||
| NM_001313483.4 | NP_001300412.1 | 5 - 6 | 1200 | 109 | ||
| NM_059306.6 | NP_491707.2 | 5 - 6 | 1037 | 109 | ||
| GenBank mRNA | EF205592.1 | - | 5 - 6 | 1025 | 109 | |
More Information
Set Membership: |
|

pgp-2
Genomic Map