Assay Details
| Gene Symbol: pgp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313483.4 | NP_001300412.1 | 8 - 9 | 2232 | 116 | |
| NM_059306.6 | NP_491707.2 | 8 - 9 | 2069 | 116 | ||
| GenBank mRNA | EF205592.1 | - | 8 - 9 | 2057 | 116 | |
More Information
Set Membership: |
|

pgp-2
Genomic Map