Assay Details
| Gene Symbol: stam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059309.8 | NP_491710.2 | 5 - 6 | 801 | 121 | |
| NM_182013.8 | NP_871813.1 | 4 - 5 | 745 | 121 | ||
More Information
Set Membership: |
|

stam-1
Genomic Map