Assay Details
| Gene Symbol: air-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059313.8 | NP_491714.1 | 5 - 6 | 897 | 83 | |
| GenBank mRNA | AF071207.1 | - | 4 - 5 | 835 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

air-2| Gene Symbol: air-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059313.8 | NP_491714.1 | 5 - 6 | 897 | 83 | |
| GenBank mRNA | AF071207.1 | - | 4 - 5 | 835 | 83 | |
Set Membership: |
|