Assay Details
Gene Symbol: set-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060371.4 | NP_492772.1 | 2 - 3 | 420 | 135 | |
NM_182049.7 | NP_871849.1 | 2 - 3 | 285 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
