Assay Details
| Gene Symbol: lam-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060374.5 | NP_492775.2 | 7 - 8 | 4283 | 146 | |
| GenBank mRNA | AF074902.1 | - | 7 - 8 | 4283 | 146 | |
More Information
Set Membership: |
|

lam-3
Genomic Map