Assay Details
| Gene Symbol: adss-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171450.3 | NP_741529.1 | 3 - 4 | 1498 | 70 | |
| NM_171451.5 | NP_741530.1 | 3 - 4 | 1316 | 70 | ||
More Information
Set Membership: |
|

adss-1
Genomic Map