Assay Details
| Gene Symbol: ttn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373034.1 | NP_001360072.1 | 45533 | 70 | ||
| NM_001380656.1 | NP_001367914.1 | 44117 | 70 | |||
| NM_001380657.1 | NP_001367912.1 | 44255 | 70 | |||
| NM_001380658.2 | NP_001367913.1 | 45395 | 70 | |||
| NM_001380659.1 | NP_001368674.1 | 28 - 29 | 23405 | 70 | ||
| NM_001380660.1 | NP_001368673.1 | 29 - 30 | 23450 | 70 | ||
More Information
Set Membership: |
|

ttn-1
Genomic Map