Assay Details
| Gene Symbol: alh-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373039.2 | NP_001360568.1 | 5 - 6 | 1196 | 77 | |
| NM_072233.7 | NP_504634.2 | 5 - 6 | 1196 | 77 | ||
| NM_171471.8 | NP_741553.1 | 5 - 6 | 1190 | 77 | ||
| NM_171472.5 | NP_741554.1 | 5 - 6 | 1196 | 77 | ||
| Gene Symbol: T05H4.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072231.7 | NP_504632.1 | 5 - 6 | 1189 | 77 | |
| Gene Symbol: atp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380669.2 | NP_001367908.1 | 5 - 6 | 1192 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380669.2, NM_072231.7 transcripts.
Set Membership: |
|

alh-4
Genomic Map