Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 14 - 15 | 2740 | 77 | |
| NM_001028947.3 | NP_001024118.1 | 11 - 12 | 2564 | 77 | ||
| NM_001028949.3 | NP_001024120.1 | 5 - 6 | 1514 | 77 | ||
| NM_001373047.2 | NP_001359916.1 | 12 - 13 | 2680 | 77 | ||
| NM_001380680.1 | NP_001367617.1 | 13 - 14 | 2720 | 77 | ||
| NM_001380681.1 | NP_001367615.1 | 13 - 14 | 2711 | 77 | ||
| NM_001380682.1 | NP_001367616.1 | 13 - 14 | 2711 | 77 | ||
| NM_171477.7 | NP_741559.3 | 14 - 15 | 2776 | 77 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map