Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 24 - 25 | 4650 | 64 | |
| NM_001028947.3 | NP_001024118.1 | 21 - 22 | 4474 | 64 | ||
| NM_001028949.3 | NP_001024120.1 | 15 - 16 | 3424 | 64 | ||
| NM_001373047.2 | NP_001359916.1 | 22 - 23 | 4590 | 64 | ||
| NM_001380680.1 | NP_001367617.1 | 23 - 24 | 4630 | 64 | ||
| NM_001380681.1 | NP_001367615.1 | 23 - 24 | 4621 | 64 | ||
| NM_171477.7 | NP_741559.3 | 24 - 25 | 4686 | 64 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map