Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 25 - 26 | 4897 | 75 | |
| NM_001028947.3 | NP_001024118.1 | 22 - 23 | 4721 | 75 | ||
| NM_001028949.3 | NP_001024120.1 | 16 - 17 | 3671 | 75 | ||
| NM_001373047.2 | NP_001359916.1 | 23 - 24 | 4837 | 75 | ||
| NM_001380680.1 | NP_001367617.1 | 24 - 25 | 4877 | 75 | ||
| NM_001380681.1 | NP_001367615.1 | 24 - 25 | 4868 | 75 | ||
| NM_171477.7 | NP_741559.3 | 25 - 26 | 4933 | 75 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map