Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380681.1 | NP_001367615.1 | 3 - 4 | 485 | 71 | |
| NM_001380682.1 | NP_001367616.1 | 3 - 4 | 485 | 71 | ||
| NM_001380683.1 | NP_001367618.1 | 3 - 4 | 485 | 71 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map