Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 8 - 9 | 1023 | 99 | |
| NM_001028947.3 | NP_001024118.1 | 5 - 6 | 847 | 99 | ||
| NM_001373047.2 | NP_001359916.1 | 6 - 7 | 963 | 99 | ||
| NM_001380680.1 | NP_001367617.1 | 7 - 8 | 1006 | 99 | ||
| NM_001380681.1 | NP_001367615.1 | 7 - 8 | 994 | 99 | ||
| NM_001380682.1 | NP_001367616.1 | 7 - 8 | 994 | 99 | ||
| NM_001380683.1 | NP_001367618.1 | 7 - 8 | 994 | 99 | ||
| NM_171477.7 | NP_741559.3 | 8 - 9 | 1059 | 99 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map