Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 10 - 11 | 1944 | 68 | |
| NM_001028947.3 | NP_001024118.1 | 7 - 8 | 1768 | 68 | ||
| NM_001028949.3 | NP_001024120.1 | 1 - 2 | 718 | 68 | ||
| NM_001373047.2 | NP_001359916.1 | 8 - 9 | 1884 | 68 | ||
| NM_001380680.1 | NP_001367617.1 | 9 - 10 | 1927 | 68 | ||
| NM_001380681.1 | NP_001367615.1 | 9 - 10 | 1915 | 68 | ||
| NM_001380682.1 | NP_001367616.1 | 9 - 10 | 1915 | 68 | ||
| NM_001380683.1 | NP_001367618.1 | 9 - 10 | 1915 | 68 | ||
| NM_171477.7 | NP_741559.3 | 10 - 11 | 1980 | 68 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map