Assay Details
| Gene Symbol: xpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380696.1 | NP_001367571.1 | 1 | 203 | 61 | |
| NM_171484.7 | NP_741567.1 | 9 | 3530 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

xpo-1| Gene Symbol: xpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380696.1 | NP_001367571.1 | 1 | 203 | 61 | |
| NM_171484.7 | NP_741567.1 | 9 | 3530 | 61 | ||
Set Membership: |
|