Assay Details
| Gene Symbol: memo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001350341.3 | NP_001337309.1 | 3 - 4 | 406 | 78 | |
| NM_001380697.2 | NP_001367639.1 | 4 - 5 | 532 | 78 | ||
| NM_001380698.1 | NP_001367638.1 | 4 - 5 | 537 | 78 | ||
More Information
Set Membership: |
|

memo-1
Genomic Map