Assay Details
| Gene Symbol: smg-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171514.6 | NP_741600.1 | 2 - 3 | 462 | 118 | |
| NM_171515.5 | NP_741601.1 | 2 - 3 | 460 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smg-4| Gene Symbol: smg-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171514.6 | NP_741600.1 | 2 - 3 | 462 | 118 | |
| NM_171515.5 | NP_741601.1 | 2 - 3 | 460 | 118 | ||
Set Membership: |
|