Assay Details
| Gene Symbol: clp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269221.3 | NP_001256150.1 | 1 - 2 | 147 | 95 | |
| NM_001269223.2 | NP_001256152.1 | 1 - 2 | 147 | 95 | ||
| NM_001383377.1 | 1 - 2 | 155 | 95 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383377.1 transcripts.
Set Membership: |
|

clp-9
Genomic Map