Assay Details
| Gene Symbol: clp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269221.3 | NP_001256150.1 | 9 - 10 | 2012 | 72 | |
| NM_001383377.1 | 9 - 10 | 2109 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

clp-9| Gene Symbol: clp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269221.3 | NP_001256150.1 | 9 - 10 | 2012 | 72 | |
| NM_001383377.1 | 9 - 10 | 2109 | 72 | |||
Set Membership: |
|