Assay Details
| Gene Symbol: crm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269281.3 | NP_001256210.1 | 5 - 6 | 1027 | 80 | |
| NM_171529.6 | NP_741616.2 | 7 - 8 | 1368 | 80 | ||
| NM_171530.6 | NP_741617.2 | 6 - 7 | 1111 | 80 | ||
| NM_171935.6 | NP_741618.2 | 6 - 7 | 1102 | 80 | ||
| NR_132530.1 | 6 - 7 | 1095 | 80 | |||
| NR_132531.1 | 6 - 7 | 1102 | 80 | |||
More Information
Set Membership: |
|

crm-1
Genomic Map