Assay Details
| Gene Symbol: crm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171530.6 | NP_741617.2 | 1 - 2 | 75 | 86 | |
| NM_171935.6 | NP_741618.2 | 1 - 2 | 66 | 86 | ||
| NR_132530.1 | 1 - 2 | 66 | 86 | |||
| NR_132531.1 | 1 - 2 | 66 | 86 | |||
More Information
Set Membership: |
|

crm-1
Genomic Map