Assay Details
| Gene Symbol: lig-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171537.5 | NP_741625.2 | 2 - 3 | 257 | 61 | |
| NM_171538.6 | NP_741626.2 | 2 - 3 | 264 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lig-1| Gene Symbol: lig-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171537.5 | NP_741625.2 | 2 - 3 | 257 | 61 | |
| NM_171538.6 | NP_741626.2 | 2 - 3 | 264 | 61 | ||
Set Membership: |
|