Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028847.4 | NP_001024018.1 | 13 - 14 | 3096 | 82 | |
| NM_001028848.4 | NP_001024019.1 | 10 - 11 | 2824 | 82 | ||
| NM_001047680.4 | NP_001041145.1 | 10 - 11 | 2706 | 82 | ||
| NM_001269630.3 | NP_001256559.1 | 10 - 11 | 2829 | 82 | ||
| NM_001269631.3 | NP_001256560.1 | 13 - 14 | 3120 | 82 | ||
| NM_001269632.3 | NP_001256561.1 | 9 - 10 | 2643 | 82 | ||
| NM_001269633.3 | NP_001256562.1 | 6 - 7 | 1635 | 82 | ||
| NM_001269634.3 | NP_001256563.1 | 5 - 6 | 1302 | 82 | ||
| NM_001307761.3 | NP_001294690.1 | 10 - 11 | 3105 | 82 | ||
| NM_001380808.2 | NP_001368670.1 | 14 - 15 | 3837 | 82 | ||
| GenBank mRNA | U40858.1 | - | 10 - 11 | 2824 | 82 | |
More Information
Set Membership: |
|

par-1
Genomic Map