Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028847.4 | NP_001024018.1 | 6 - 7 | 1355 | 108 | |
| NM_001028848.4 | NP_001024019.1 | 4 - 5 | 1227 | 108 | ||
| NM_001047680.4 | NP_001041145.1 | 3 - 4 | 965 | 108 | ||
| NM_001129495.4 | NP_001122967.2 | 3 - 4 | 1163 | 108 | ||
| NM_001269630.3 | NP_001256559.1 | 3 - 4 | 1088 | 108 | ||
| NM_001269631.3 | NP_001256560.1 | 6 - 7 | 1355 | 108 | ||
| NM_001269632.3 | NP_001256561.1 | 2 - 3 | 902 | 108 | ||
| NM_001307761.3 | NP_001294690.1 | 3 - 4 | 1364 | 108 | ||
| NM_001373155.1 | NP_001359744.1 | 3 - 4 | 1163 | 108 | ||
| NM_001380808.2 | NP_001368670.1 | 6 - 7 | 1652 | 108 | ||
| GenBank mRNA | U40858.1 | - | 4 - 5 | 1227 | 108 | |
More Information
Set Membership: |
|

par-1
Genomic Map