Assay Details
| Gene Symbol: slo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029090.5 | NP_001024261.1 | 12 - 13 | 2118 | 63 | |
| NM_001269872.3 | NP_001256801.1 | 13 - 14 | 2118 | 63 | ||
| NM_001269873.3 | NP_001256802.1 | 12 - 13 | 2094 | 63 | ||
| NM_001269874.3 | NP_001256803.1 | 12 - 13 | 2094 | 63 | ||
| NM_001269875.4 | NP_001256804.1 | 12 - 13 | 2118 | 63 | ||
| NM_001269878.3 | NP_001256807.1 | 12 - 13 | 2094 | 63 | ||
| NM_001269879.4 | NP_001256808.1 | 12 - 13 | 2139 | 63 | ||
| NM_001269880.3 | NP_001256809.1 | 12 - 13 | 2073 | 63 | ||
| NM_001269881.3 | NP_001256810.1 | 12 - 13 | 2073 | 63 | ||
| GenBank mRNA | AF431893.1 | - | 12 - 13 | 2073 | 63 | |
| JQ582463.1 | - | 12 - 13 | 2094 | 63 | ||
| JQ606809.1 | - | 12 - 13 | 2073 | 63 | ||
| JQ606812.1 | - | 12 - 13 | 2073 | 63 | ||
More Information
Set Membership: |
|

slo-1
Genomic Map