Assay Details
| Gene Symbol: slo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029089.3 | NP_001024260.1 | 12 - 13 | 2183 | 82 | |
| NM_001029090.5 | NP_001024261.1 | 13 - 14 | 2222 | 82 | ||
| NM_001269872.3 | NP_001256801.1 | 14 - 15 | 2222 | 82 | ||
| NM_001269874.3 | NP_001256803.1 | 13 - 14 | 2198 | 82 | ||
| NM_001269877.3 | NP_001256806.1 | 12 - 13 | 2138 | 82 | ||
| NM_001269879.4 | NP_001256808.1 | 13 - 14 | 2243 | 82 | ||
| NM_001269881.3 | NP_001256810.1 | 13 - 14 | 2177 | 82 | ||
| GenBank mRNA | AF431892.1 | - | 12 - 13 | 2138 | 82 | |
| AF431893.1 | - | 13 - 14 | 2177 | 82 | ||
| JQ606810.1 | - | 12 - 13 | 2138 | 82 | ||
| JQ606812.1 | - | 13 - 14 | 2177 | 82 | ||
More Information
Set Membership: |
|

slo-1
Genomic Map