Assay Details
| Gene Symbol: slo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029088.2 | NP_001024259.1 | 3 - 4 | 517 | 100 | |
| NM_001029089.3 | NP_001024260.1 | 3 - 4 | 523 | 100 | ||
| NM_001029090.5 | NP_001024261.1 | 3 - 4 | 523 | 100 | ||
| NM_001269872.3 | NP_001256801.1 | 4 - 5 | 523 | 100 | ||
| NM_001269873.3 | NP_001256802.1 | 3 - 4 | 478 | 100 | ||
| NM_001269874.3 | NP_001256803.1 | 3 - 4 | 478 | 100 | ||
| NM_001269875.4 | NP_001256804.1 | 3 - 4 | 523 | 100 | ||
| NM_001269876.3 | NP_001256805.1 | 3 - 4 | 478 | 100 | ||
| NM_001269877.3 | NP_001256806.1 | 3 - 4 | 478 | 100 | ||
| NM_001269878.3 | NP_001256807.1 | 3 - 4 | 478 | 100 | ||
| NM_001269879.4 | NP_001256808.1 | 3 - 4 | 523 | 100 | ||
| NM_001269880.3 | NP_001256809.1 | 3 - 4 | 478 | 100 | ||
| NM_001269881.3 | NP_001256810.1 | 3 - 4 | 478 | 100 | ||
| GenBank mRNA | AF431891.1 | - | 3 - 4 | 478 | 100 | |
| AF431892.1 | - | 3 - 4 | 478 | 100 | ||
| AF431893.1 | - | 3 - 4 | 478 | 100 | ||
| JQ582463.1 | - | 3 - 4 | 478 | 100 | ||
| JQ606809.1 | - | 3 - 4 | 478 | 100 | ||
| JQ606810.1 | - | 3 - 4 | 478 | 100 | ||
| JQ606812.1 | - | 3 - 4 | 478 | 100 | ||
More Information
Set Membership: |
|

slo-1
Genomic Map