Assay Details
| Gene Symbol: zhp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171592.5 | NP_741682.1 | 4 - 5 | 1429 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zhp-4| Gene Symbol: zhp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171592.5 | NP_741682.1 | 4 - 5 | 1429 | 112 | |
Set Membership: |
|