Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270046.2 | NP_001256975.1 | 3 - 4 | 863 | 78 | |
| NM_001374936.2 | NP_001362085.1 | 16 - 17 | 2962 | 78 | ||
| NM_001374938.1 | NP_001362002.1 | 17 - 18 | 3035 | 78 | ||
| NM_001380893.1 | NP_001367765.1 | 13 - 14 | 2743 | 78 | ||
| NM_001383522.1 | 3 - 4 | 850 | 78 | |||
More Information
Set Membership: |
|

ifc-2
Genomic Map