Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270046.2 | NP_001256975.1 | 4 - 5 | 1021 | 91 | |
| NM_001374936.2 | NP_001362085.1 | 17 - 18 | 3120 | 91 | ||
| NM_001374938.1 | NP_001362002.1 | 18 - 19 | 3193 | 91 | ||
| NM_001380893.1 | NP_001367765.1 | 14 - 15 | 2901 | 91 | ||
| NM_001383522.1 | 4 - 5 | 1008 | 91 | |||
More Information
Set Membership: |
|

ifc-2
Genomic Map