Assay Details
| Gene Symbol: ifc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270046.2 | NP_001256975.1 | 5 - 6 | 1191 | 74 | |
| NM_001374936.2 | NP_001362085.1 | 18 - 19 | 3290 | 74 | ||
| NM_001374938.1 | NP_001362002.1 | 19 - 20 | 3363 | 74 | ||
| NM_001380893.1 | NP_001367765.1 | 15 - 16 | 3071 | 74 | ||
| NM_001383522.1 | 5 - 6 | 1178 | 74 | |||
More Information
Set Membership: |
|

ifc-2
Genomic Map